Assay Details
Gene Symbol: Srpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108243.2 | NP_001101713.1 | 9 - 10 | 1219 | 73 | |
GenBank mRNA | BC168975.1 | - | 9 - 10 | 1219 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
