Assay Details
Gene Symbol: Nt5c1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011982.1 | NP_001011982.1 | 1 - 2 | 121 | 151 | |
XM_006239888.2 | XP_006239950.1 | 1 - 2 | 149 | 151 | ||
XM_006239889.2 | XP_006239951.1 | 1 - 2 | 149 | 151 | ||
XM_006239891.2 | XP_006239953.1 | 1 - 2 | 149 | 151 | ||
GenBank mRNA | BC079437.1 | - | 1 - 2 | 121 | 151 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC079437.1, NM_001011982.1, XM_006239888.2, XM_006239889.2, XM_006239891.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
