Assay Details
Gene Symbol: Nt5c1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011982.1 | NP_001011982.1 | 7 - 8 | 1296 | 83 | |
XM_006239888.2 | XP_006239950.1 | 8 - 9 | 1498 | 83 | ||
XM_006239889.2 | XP_006239951.1 | 8 - 9 | 1495 | 83 | ||
XM_006239890.2 | XP_006239952.1 | 8 - 9 | 1375 | 83 | ||
XM_006239891.2 | XP_006239953.1 | 7 - 8 | 1321 | 83 | ||
XM_006239892.2 | XP_006239954.1 | 7 - 8 | 1332 | 83 | ||
GenBank mRNA | BC079437.1 | - | 7 - 8 | 1296 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
