Assay Details
Gene Symbol: Memo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029917.1 | NP_001025088.1 | 3 - 4 | 354 | 74 | |
GenBank mRNA | BC098061.1 | - | 3 - 4 | 354 | 74 | |
FQ215386.1 | - | 1 - 2 | 106 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
