Assay Details
Gene Symbol: Memo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029917.1 | NP_001025088.1 | 6 - 7 | 677 | 171 | |
GenBank mRNA | BC098061.1 | - | 6 - 7 | 677 | 171 | |
FQ215386.1 | - | 4 - 5 | 429 | 171 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P443412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80818 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P443412 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P443412 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P443412 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P443412 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51198 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100760 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44070 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22093 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10882927 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10882928 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10882921 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10882925 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10882926 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10882922 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10882923 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10882924 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10882920 | - | - |
Rat Genome 230 2.0 Array | 1376083_at | - | - |
Rat Genome 230 2.0 Array | 1389312_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389312_at | - | - |
Rat Genome 230 2.0 Array | 1376083_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389312_at | - | - |
Affymetrix HT Rat Focus Array | 1389312_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600001410.rn.1 | TC0600000222.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600001412.rn.1 | TC0600000222.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600001411.rn.1 | TC0600000222.rn.1 | RTA TC |
Rat Expression Set 230 | 1376083_PM_at | - | - |
Rat Expression Set 230 | 1376083_at | - | - |
Rat Expression Set 230 | 1389312_PM_at | - | - |
Rat Expression Set 230 | 1389312_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10882920 | - | - |
Rat Genome U34 Set | rc_AI045595_at | - | - |
Rat Genome U34 Set | rc_AA925064_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351135 | - | - |
