Assay Details
Gene Symbol: Cenpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106711.1 | NP_001100181.1 | 2 - 3 | 286 | 79 | |
XM_006239798.2 | XP_006239860.1 | 2 - 3 | 409 | 79 | ||
GenBank mRNA | BC166997.1 | - | 2 - 3 | 327 | 8365241 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
