Assay Details
Gene Symbol: Pnpla8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750150.3 | XP_003750198.1 | 3 - 4 | 1406 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
