Assay Details
Gene Symbol: Pnpla8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750150.3 | XP_003750198.1 | 6 - 7 | 1829 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
