Assay Details
Gene Symbol: Cfl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108982.2 | NP_001102452.2 | 1 - 2 | 136 | 156 | |
GenBank mRNA | FQ228820.1 | - | 1 - 2 | 139 | 156 | |
FQ228857.1 | - | 1 - 2 | 143 | 156 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108982.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
