Assay Details
Gene Symbol: Cbll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108018.1 | NP_001101488.1 | 1 - 2 | 90 | 65 | |
XM_006239986.2 | XP_006240048.1 | 1 - 2 | 94 | 65 | ||
XM_006239987.2 | XP_006240049.1 | 1 - 2 | 95 | 65 | ||
XM_006239988.2 | XP_006240050.1 | 1 - 2 | 95 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108018.1, XM_006239986.2, XM_006239987.2, XM_006239988.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
