Assay Details
Gene Symbol: Cbll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108018.1 | NP_001101488.1 | 4 - 5 | 429 | 139 | |
XM_006239986.2 | XP_006240048.1 | 4 - 5 | 442 | 139 | ||
XM_006239987.2 | XP_006240049.1 | 4 - 5 | 440 | 139 | ||
XM_006239988.2 | XP_006240050.1 | 4 - 5 | 431 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
