Assay Details
Gene Symbol: Epcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138541.1 | NP_612550.1 | 6 - 7 | 723 | 99 | |
GenBank mRNA | AJ001044.1 | - | 6 - 7 | 778 | 99 | |
BC061787.1 | - | 6 - 7 | 727 | 99 | ||
BC072691.1 | - | 6 - 7 | 747 | 99 | ||
FQ229377.1 | - | 1 - 2 | 111 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
