Assay Details
Gene Symbol: Fam49a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106718.2 | NP_001100188.1 | 2 - 3 | 179 | 85 | |
XM_008764584.1 | XP_008762806.1 | 2 - 3 | 728 | 85 | ||
GenBank mRNA | BC158700.1 | - | 2 - 3 | 179 | 85 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC158700.1, NM_001106718.2, XM_008764584.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
