Assay Details
Gene Symbol: Fam49a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106718.2 | NP_001100188.1 | 3 - 4 | 266 | 125 | |
XM_008764584.1 | XP_008762806.1 | 3 - 4 | 815 | 125 | ||
XM_008764585.1 | XP_008762807.1 | 2 - 3 | 168 | 125 | ||
GenBank mRNA | BC158700.1 | - | 3 - 4 | 266 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
