Assay Details
Gene Symbol: Fam49a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106718.2 | NP_001100188.1 | 6 - 7 | 633 | 156 | |
XM_008764584.1 | XP_008762806.1 | 6 - 7 | 1182 | 156 | ||
XM_008764585.1 | XP_008762807.1 | 5 - 6 | 535 | 156 | ||
GenBank mRNA | BC158700.1 | - | 6 - 7 | 633 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
