Assay Details
Gene Symbol: Prkar2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030020.1 | NP_001025191.1 | 6 - 7 | 745 | 109 | |
GenBank mRNA | BC099223.1 | - | 5 - 6 | 488 | 7388087 | 109 |
M12492.1 | - | 6 - 7 | 739 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
