Assay Details
Gene Symbol: Dcaf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100718.1 | NP_001094188.1 | 6 - 7 | 892 | 117 | |
XM_006240279.2 | XP_006240341.1 | 6 - 7 | 694 | 117 | ||
XM_008764782.1 | XP_008763004.1 | 6 - 7 | 747 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
