Assay Details
Gene Symbol: Atp6v1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014199.1 | NP_001014221.1 | 7 - 8 | 697 | 119 | |
GenBank mRNA | AY594319.1 | - | 6 - 7 | 622 | 119 | |
BC079083.1 | - | 7 - 8 | 697 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
