Assay Details
Gene Symbol: Pdia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004442.1 | NP_001004442.1 | 12 - 13 | 1283 | 96 | |
GenBank mRNA | BC082063.1 | - | 12 - 13 | 1283 | 96 | |
FQ229584.1 | - | 7 - 8 | 741 | 96 | ||
X79328.1 | - | 11 - 12 | 1223 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
