Assay Details
Gene Symbol: Pdia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004442.1 | NP_001004442.1 | 6 - 7 | 618 | 92 | |
GenBank mRNA | BC082063.1 | - | 6 - 7 | 618 | 92 | |
FQ229584.1 | - | 1 - 2 | 76 | 92 | ||
X79328.1 | - | 5 - 6 | 558 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
