Assay Details
Gene Symbol: Rbks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108703.1 | NP_001102173.1 | 4 - 5 | 386 | 117 | |
XM_006239806.2 | XP_006239868.1 | 4 - 5 | 432 | 117 | ||
XM_008764520.1 | XP_008762742.1 | 4 - 5 | 880 | 117 | ||
XM_008764521.1 | XP_008762743.1 | 4 - 5 | 432 | 117 | ||
GenBank mRNA | FQ209797.1 | - | 4 - 5 | 368 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
