Assay Details
Gene Symbol: Rbks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108703.1 | NP_001102173.1 | 6 - 7 | 639 | 55 | |
XM_006239805.2 | XP_006239867.1 | 5 - 6 | 520 | 55 | ||
XM_008764520.1 | XP_008762742.1 | 6 - 7 | 1133 | 55 | ||
XM_008764521.1 | XP_008762743.1 | 6 - 7 | 685 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P122457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48358 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P122457 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P122457 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P122457 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P568034 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 80967 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 55410 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103788 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10883084 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883082 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883080 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883081 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883078 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883079 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883077 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883083 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10883076 | - | - |
Rat Genome 230 2.0 Array | 1375093_at | - | - |
Rat Genome 230 2.0 Array | 1375093_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375093_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375093_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600001649.rn.1 | TC0600000269.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600001650.rn.1 | TC0600000269.rn.1 | RTA TC |
Rat Expression Set 230 | 1375093_at | - | - |
Rat Expression Set 230 | 1375093_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883076 | - | - |
Rat Genome U34 Set | rc_AI236916_s_at | - | - |
Rat Genome U34 Set | rc_AI236089_at | - | - |
Rat Genome U34 Set | rc_AI175440_at | - | - |
Rat Genome U34 Set | rc_AI233177_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21893086 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374781 | - | - |
