Assay Details
Gene Symbol: Agr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106725.1 | NP_001100195.1 | 6 - 7 | 623 | 75 | |
XM_006240041.2 | XP_006240103.1 | 6 - 7 | 625 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
