Assay Details
Gene Symbol: Npas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764673.1 | XP_008762895.1 | 3 - 4 | 734 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
