Assay Details
Gene Symbol: Tcea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025735.1 | NP_001020906.1 | 1 - 2 | 147 | 136 | |
XM_006237816.2 | XP_006237878.1 | 1 - 2 | 208 | 136 | ||
GenBank mRNA | BC099141.1 | - | 1 - 2 | 147 | 136 | |
FQ227501.1 | - | 1 - 2 | 119 | 136 |
Gene Symbol: LOC498453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251720.2 | XP_006251782.1 | 1 | 201 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
