Assay Details
Gene Symbol: Tcea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025735.1 | NP_001020906.1 | 2 - 3 | 216 | 96 | |
XM_006237816.2 | XP_006237878.1 | 2 - 3 | 277 | 96 | ||
XM_008763546.1 | XP_008761768.1 | 2 - 3 | 104 | 96 | ||
GenBank mRNA | BC099141.1 | - | 2 - 3 | 216 | 96 | |
FQ227501.1 | - | 2 - 3 | 188 | 96 |
Gene Symbol: LOC498453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251720.2 | XP_006251782.1 | 1 | 270 | 96 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_008763546.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
