Assay Details
Gene Symbol: Lypla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013006.1 | NP_037138.1 | 6 - 7 | 432 | 147 | |
XM_008763522.1 | XP_008761744.1 | 5 - 6 | 424 | 147 | ||
GenBank mRNA | BC085750.1 | - | 6 - 7 | 370 | 147 | |
D63885.1 | - | 6 - 7 | 432 | 147 | ||
U97146.1 | - | 6 - 7 | 460 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
