Assay Details
Gene Symbol: Apool | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014105.1 | NP_001014127.1 | 6 - 7 | 624 | 108 | |
XM_006257191.2 | XP_006257253.1 | 6 - 7 | 563 | 108 | ||
GenBank mRNA | BC086434.1 | - | 6 - 7 | 624 | 108 | |
FQ219134.1 | - | 6 - 7 | 514 | 108 | ||
FQ235331.1 | - | 6 - 7 | 510 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
