Assay Details
Gene Symbol: Apool | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014105.1 | NP_001014127.1 | 7 - 8 | 745 | 94 | |
XM_006257191.2 | XP_006257253.1 | 7 - 8 | 684 | 94 | ||
GenBank mRNA | BC086434.1 | - | 7 - 8 | 745 | 94 | |
FQ219134.1 | - | 7 - 8 | 635 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P589570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79087 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P589570 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P589570 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P589570 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P589570 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17458 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25871 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49279 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1425 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66490 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1251 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10934707 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934704 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934709 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934706 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934708 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934701 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934702 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934703 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934705 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10934700 | - | - |
Rat Genome 230 2.0 Array | 1389148_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389148_at | - | - |
Affymetrix HT Rat Focus Array | 1389148_at | - | - |
Affymetrix HT Rat Focus Array | 1389148_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00004801.rn.1 | TC0X00000774.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00004800.rn.1 | TC0X00000774.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00004803.rn.1 | TC0X00000774.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00004804.rn.1 | TC0X00000774.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00004802.rn.1 | TC0X00000774.rn.1 | RTA TC |
Rat Expression Set 230 | 1389148_PM_at | - | - |
Rat Expression Set 230 | 1389148_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10934700 | - | - |
Rat Genome U34 Set | rc_AI177050_at | - | - |
Rat Genome U34 Set | rc_AI104883_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20836779 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369989 | - | - |
