Assay Details
Gene Symbol: Mrpl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107567.1 | NP_001101037.1 | 3 - 4 | 411 | 113 | |
XM_008760120.1 | XP_008758342.1 | 3 - 4 | 313 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
