Assay Details
Gene Symbol: Slc25a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014027.1 | NP_001014049.1 | 8 - 9 | 749 | 94 | |
XM_006230557.2 | XP_006230619.1 | 10 - 11 | 1088 | 94 | ||
XM_008759993.1 | XP_008758215.1 | 9 - 10 | 1055 | 94 | ||
XM_008759994.1 | XP_008758216.1 | 9 - 10 | 1061 | 94 | ||
XM_008759995.1 | XP_008758217.1 | 9 - 10 | 1013 | 94 | ||
XM_008759996.1 | XP_008758218.1 | 9 - 10 | 1616 | 94 | ||
GenBank mRNA | BC090010.1 | - | 8 - 9 | 749 | 7324394 | 94 |
Gene Symbol: LOC100911440 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749008.3 | XP_003749056.1 | 1088 | 94 | ||
XM_008760026.1 | XP_008758248.1 | 1055 | 94 | |||
XM_008760027.1 | XP_008758249.1 | 1061 | 94 | |||
XM_008760028.1 | XP_008758250.1 | 1013 | 94 | |||
XM_008760029.1 | XP_008758251.1 | 1616 | 94 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
