Assay Details
Gene Symbol: Cmss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013866.1 | NP_001013888.1 | 4 - 5 | 471 | 142 | |
GenBank mRNA | BC089826.1 | - | 4 - 5 | 471 | 142 | |
FQ214930.1 | - | 4 - 5 | 477 | 142 | ||
FQ219915.1 | - | 4 - 5 | 472 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
