Assay Details
Gene Symbol: Senp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768807.1 | XP_008767029.1 | 4 - 5 | 1978 | 72 | |
XM_008768808.1 | XP_008767030.1 | 4 - 5 | 1974 | 72 | ||
XM_221369.8 | XP_221369.2 | 4 - 5 | 1977 | 72 |
Gene Symbol: LOC103689978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248475.2 | XP_006248537.1 | 4 - 5 | 1977 | 72 | |
XM_008768779.1 | XP_008767001.1 | 4 - 5 | 1978 | 72 | ||
XM_008768780.1 | XP_008767002.1 | 4 - 5 | 1974 | 72 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
