Assay Details
Gene Symbol: Tmem39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013865.1 | NP_001013887.1 | 1 - 2 | 278 | 59 | |
XM_008768717.1 | XP_008766939.1 | 1 - 2 | 317 | 59 | ||
GenBank mRNA | BC085845.1 | - | 1 - 2 | 278 | 59 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC085845.1, NM_001013865.1, XM_008768717.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
