Assay Details
Gene Symbol: Osbpl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107090.2 | NP_001100560.1 | 9 - 10 | 2122 | 78 | |
XM_006248450.2 | XP_006248512.1 | 8 - 9 | 2177 | 78 | ||
XM_006248451.2 | XP_006248513.1 | 9 - 10 | 1753 | 78 | ||
XM_006248452.2 | XP_006248514.1 | 9 - 10 | 1634 | 78 | ||
XM_008768726.1 | XP_008766948.1 | 9 - 10 | 1903 | 78 | ||
GenBank mRNA | BC168952.1 | - | 9 - 10 | 2122 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
