Assay Details
Gene Symbol: Slc25a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257534.2 | XP_006257596.1 | 2 - 3 | 1194 | 105 | |
XM_008773531.1 | XP_008771753.1 | 3 - 4 | 151 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006257534.2, XM_008773531.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
