Assay Details
Gene Symbol: Plaur | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017350.1 | NP_059046.1 | 4 - 5 | 484 | 59 | |
NM_134352.1 | NP_599179.1 | 4 - 5 | 489 | 59 | ||
GenBank mRNA | AF007789.1 | - | 4 - 5 | 476 | 59 | |
BC127499.1 | - | 4 - 5 | 484 | 7380245 | 59 | |
FQ220340.1 | - | 4 - 5 | 497 | 59 | ||
X71898.1 | - | 4 - 5 | 489 | 59 | ||
X71899.1 | - | 4 - 5 | 484 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
