Assay Details
Gene Symbol: Gas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127504.1 | NP_001120976.1 | 3 - 4 | 513 | 92 | |
XM_006229270.2 | XP_006229332.1 | 4 - 5 | 978 | 92 | ||
XM_006229271.2 | XP_006229333.1 | 3 - 4 | 432 | 92 | ||
XM_006229273.2 | XP_006229335.1 | 4 - 5 | 770 | 92 | ||
XM_006229274.2 | XP_006229336.1 | 4 - 5 | 1297 | 92 | ||
XM_008759407.1 | XP_008757629.1 | 3 - 4 | 511 | 92 | ||
XM_008759408.1 | XP_008757630.1 | 3 - 4 | 432 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
