Assay Details
Gene Symbol: Gpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207592.1 | NP_997475.1 | 5 - 6 | 555 | 146 | |
GenBank mRNA | BC062005.1 | - | 5 - 6 | 555 | 5599776 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
