Assay Details
Gene Symbol: Akt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017093.1 | NP_058789.1 | 12 - 13 | 1549 | 64 | |
XM_008759108.1 | XP_008757330.1 | 13 - 14 | 3948 | 64 | ||
XM_008759109.1 | XP_008757331.1 | 12 - 13 | 1536 | 64 | ||
XM_008759110.1 | XP_008757332.1 | 12 - 13 | 3813 | 64 | ||
XM_008759111.1 | XP_008757333.1 | 12 - 13 | 1659 | 64 | ||
GenBank mRNA | D30041.1 | - | 12 - 13 | 1549 | 64 | |
DQ198085.1 | - | 11 - 12 | 1268 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
