Assay Details
Gene Symbol: Tgfb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021578.2 | NP_067589.1 | 3 - 4 | 779 | 78 | |
GenBank mRNA | AY550025.1 | - | 3 - 4 | 634 | 78 | |
BC076380.1 | - | 3 - 4 | 779 | 7107411 | 78 | |
X52498.1 | - | 3 - 4 | 1046 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
