Assay Details
Gene Symbol: Rhpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107505.1 | NP_001100975.1 | 13 - 14 | 1676 | 80 | |
XM_006228887.2 | XP_006228949.1 | 13 - 14 | 1723 | 80 | ||
XM_008759160.1 | XP_008757382.1 | 13 - 14 | 1723 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
