Assay Details
Gene Symbol: Slc7a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053726.2 | NP_446178.2 | 4 - 5 | 706 | 62 | |
XM_006228873.2 | XP_006228935.1 | 4 - 5 | 784 | 62 | ||
XM_006228874.2 | XP_006228936.1 | 4 - 5 | 784 | 62 | ||
GenBank mRNA | AB126813.1 | - | 4 - 5 | 706 | 62 | |
BC127467.1 | - | 4 - 5 | 677 | 7313174 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
