Assay Details
Gene Symbol: Fxyd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270688.1 | NP_001257617.1 | 5 - 6 | 342 | 66 | |
NM_001270689.1 | NP_001257618.1 | 5 - 6 | 370 | 66 | ||
NM_021909.2 | NP_068709.2 | 5 - 6 | 446 | 66 | ||
GenBank mRNA | DV726876.1 | - | 1 - 2 | 112 | 66 | |
FM122563.1 | - | 5 - 6 | 320 | 66 | ||
FQ220058.1 | - | 5 - 6 | 370 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
