Assay Details
Gene Symbol: Fxyd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270688.1 | NP_001257617.1 | 8 - 9 | 521 | 84 | |
NM_001270689.1 | NP_001257618.1 | 8 - 9 | 549 | 84 | ||
NM_021909.2 | NP_068709.2 | 8 - 9 | 625 | 84 | ||
GenBank mRNA | AI407979.1 | - | 1 - 2 | 84 | ||
BC060571.1 | - | 8 - 9 | 513 | 84 | ||
DV726876.1 | - | 4 - 5 | 291 | 84 | ||
FM122563.1 | - | 8 - 9 | 499 | 84 | ||
FQ220058.1 | - | 8 - 9 | 549 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
