Assay Details
Gene Symbol: Fxyd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270688.1 | NP_001257617.1 | 2 - 3 | 105 | 72 | |
NM_001270689.1 | NP_001257618.1 | 2 - 3 | 133 | 72 | ||
NM_021909.2 | NP_068709.2 | 2 - 3 | 209 | 72 | ||
GenBank mRNA | BC060571.1 | - | 2 - 3 | 97 | 6891214 | 72 |
FM122563.1 | - | 2 - 3 | 83 | 72 | ||
FQ220058.1 | - | 2 - 3 | 133 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
