Assay Details
Gene Symbol: Scn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271045.1 | NP_001257974.1 | 1 - 2 | 178 | 87 | |
NM_017288.2 | NP_058984.1 | 1 - 2 | 259 | 87 | ||
GenBank mRNA | AF182949.1 | - | 1 - 2 | 39 | 87 | |
BC094523.1 | - | 1 - 2 | 178 | 7316686 | 87 | |
M91808.1 | - | 1 - 2 | 258 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
