Assay Details
Gene Symbol: Ceacam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033860.1 | NP_001029032.1 | 5 - 6 | 1290 | 81 | |
NM_001033862.1 | NP_001029034.1 | 5 - 6 | 1290 | 81 | ||
GenBank mRNA | BC061740.1 | - | 5 - 6 | 1276 | 81 | |
J04963.1 | - | 5 - 6 | 1297 | 81 | ||
M92848.1 | - | 5 - 6 | 1290 | 81 | ||
X71122.1 | - | 5 - 6 | 1233 | 81 | ||
X91137.1 | - | 5 - 6 | 1252 | 81 | ||
Z12019.1 | - | 5 - 6 | 1253 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
