Assay Details
Gene Symbol: Fuz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037646.1 | NP_001032735.1 | 2 - 3 | 376 | 73 | |
XM_006229059.2 | XP_006229121.1 | 2 - 3 | 415 | 73 | ||
XM_006229060.2 | XP_006229122.1 | 2 - 3 | 415 | 73 | ||
XM_008759373.1 | XP_008757595.1 | 2 - 3 | 416 | 73 | ||
XM_008759374.1 | XP_008757596.1 | 2 - 3 | 416 | 73 | ||
GenBank mRNA | BC107809.1 | - | 2 - 3 | 376 | 73 | |
FQ228777.1 | - | 2 - 3 | 361 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
