Assay Details
Gene Symbol: Gapdhs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023964.1 | NP_076454.1 | 9 - 10 | 1131 | 94 | |
XM_006228790.2 | XP_006228852.1 | 9 - 10 | 1501 | 94 | ||
XM_006228791.2 | XP_006228853.1 | 9 - 10 | 1362 | 94 | ||
GenBank mRNA | AJ297631.1 | - | 9 - 10 | 1131 | 94 | |
BC161847.1 | - | 9 - 10 | 1153 | 7112898 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
